Skip to Content
Merck
All Photos(1)

Key Documents

LL/2(LLc1)

90020104, mouse lung, Not specified

Synonym(s):

LLC1 Cells, Lewis lung carcinoma line 1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

LL/2(LLc1), NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 90020104, mouse lung, Not specified, 90020104

biological source

mouse lung

packaging

tube of 5 μg 90020104-DNA-5UG
pkg of vial of cells 90020104-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C57BL Lewis lung carcinoma

Cell Line Description

Derived from the lung of a C57BL mouse implanted with a primary Lewis Lung carcinoma. The cells can be used as a model for metastasis and for studying the effects of chemotherapeutic agents.

Application

Tumourigenicity, Metastasis and chemotherapeutic agent studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using mechanical technique (aspiration); 5% CO2; 37°C. Cells detach as aggregates and are therefore difficult to count.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service