Recommended Products
grade
certified reference material
Quality Level
CofA
current certificate can be downloaded
manufacturer/tradename
RTC SQC-013
technique(s)
ion chromatography: suitable
matrix
soil
application(s)
environmental
food and beverages
format
matrix material
storage temp.
2-30°C
General description
Other Notes
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service