HLA003
A549 HLA Panel Cell Lines
human lung (Carcinoma), Epithelial
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Product Name
A549 HLA Panel Cell Lines,
biological source
human lung (Carcinoma)
growth mode
Adherent
morphology
Epithelial
shipped in
dry ice
storage temp.
−196°C
General description
The STR profile of these cell lines match that of its parental cell line - ECACC catalog number 86012804.
This product is a panel of eleven (11) genetically modified cell lines targeting MHC Class I HLA molecules in an A549 lung carcinoma parental cell line background. The full HLA panel consists of a Beta-2 microglobulin (β2M) knockout cell line and an additional ten (10) monoallelic HLA expression cell lines.
CompoZr® Zinc Finger Nuclease (ZFN) technology was used to generate the β2M knockout cell line, which does not express endogenous cell surface MHC Class I HLA molecules (HLA-A or HLA-B) as measured by fluorescence-activated cell sorting (FACS).
MISSION® lentivirus was used to generate the ten (10) monoallelic HLA cell lines, which express individual HLA-A or HLA-B subtypes on the cell surface via a β2M:HLA fusion protein as described in Nature Biotechnology 35, 765-772, (2017) and as demonstrated via FACS analysis.
This product is a panel of eleven (11) genetically modified cell lines targeting MHC Class I HLA molecules in an A549 lung carcinoma parental cell line background. The full HLA panel consists of a Beta-2 microglobulin (β2M) knockout cell line and an additional ten (10) monoallelic HLA expression cell lines.
CompoZr® Zinc Finger Nuclease (ZFN) technology was used to generate the β2M knockout cell line, which does not express endogenous cell surface MHC Class I HLA molecules (HLA-A or HLA-B) as measured by fluorescence-activated cell sorting (FACS).
MISSION® lentivirus was used to generate the ten (10) monoallelic HLA cell lines, which express individual HLA-A or HLA-B subtypes on the cell surface via a β2M:HLA fusion protein as described in Nature Biotechnology 35, 765-772, (2017) and as demonstrated via FACS analysis.
Cell Line Description
Derived from a 58 year old Caucasian male. The cells can synthesise lecithin utilising the cytidine diphosphocholine pathway. Occasional cells may also contain inclusion bodies although they are not known to carry any human pathogen.
Features and Benefits
Unlike wild type A549 cells, which express multiple HLA subtypes, the A549 HLA panel cell lines express individual MHC Class I HLA subtypes. These monoallelic HLA cell lines can be used to study peptide binding and presentation in the context of individual HLA subtypes. The panel can also be used to develop cell-based assays to evaluate candidate cancer immunotherapies and identify tumor-specific neoantigens.
Components
HLA003A-1VL: A549 B2M KO Cells
HLA003B-1VL: A549 HLA-A*02:01 Cells
HLA003C-1VL: A549 HLA-A*01:01 Cells
HLA003D-1VL: A549 HLA-A*03:01 Cells
HLA003E-1VL: A549 HLA-A*11:01 Cells
HLA003F-1VL: A549 HLA-A*24:02 Cells
HLA003G-1VL: A549 HLA-B*15:10 Cells
HLA003H-1VL: A549 HLA-B*07:02 Cells
HLA003I-1VL: A549 HLA-B*08:01 Cells
HLA003J-1VL: A549 HLA-B*35:01 Cells
HLA003K-1VL: A549 HLA-B*40:01 Cells
HLA003B-1VL: A549 HLA-A*02:01 Cells
HLA003C-1VL: A549 HLA-A*01:01 Cells
HLA003D-1VL: A549 HLA-A*03:01 Cells
HLA003E-1VL: A549 HLA-A*11:01 Cells
HLA003F-1VL: A549 HLA-A*24:02 Cells
HLA003G-1VL: A549 HLA-B*15:10 Cells
HLA003H-1VL: A549 HLA-B*07:02 Cells
HLA003I-1VL: A549 HLA-B*08:01 Cells
HLA003J-1VL: A549 HLA-B*35:01 Cells
HLA003K-1VL: A549 HLA-B*40:01 Cells
Quality
Tested for Mycoplasma, bacterial and fungal content, post-freeze viability, and short terminal repeat (STR) analysis for cell line identification.
Legal Information
CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service