Skip to Content
Merck
All Photos(1)

Key Documents

74711

Sigma-Aldrich

Ochratoxin A

purum, ≥98.0% (TLC), from Aspergillus ochraceus

Synonym(s):

N-[(3R)-(5-Chloro-8-hydroxy-3-methyl-1-oxo-7-isochromanyl)carbonyl]-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18ClNO6
CAS Number:
Molecular Weight:
403.81
Beilstein:
1301486
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

Aspergillus ochraceus

grade

purum

Assay

≥98.0% (TLC)

form

powder

Mode of action

enzyme | inhibits
protein synthesis | interferes

storage temp.

2-8°C

SMILES string

C[C@@H]1Cc2c(Cl)cc(c(O)c2C(=O)O1)C(=O)N[C@@H](Cc3ccccc3)C(O)=O

InChI

1S/C20H18ClNO6/c1-10-7-12-14(21)9-13(17(23)16(12)20(27)28-10)18(24)22-15(19(25)26)8-11-5-3-2-4-6-11/h2-6,9-10,15,23H,7-8H2,1H3,(H,22,24)(H,25,26)/t10-,15+/m1/s1

InChI key

RWQKHEORZBHNRI-BMIGLBTASA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Inhibitor of mitochondrial transport systems; Mode of action.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Roschenthaler et al.
J. Toxicol.-Toxin Reviews, 3, 53-53 (1984)
Ochratoxin A, an inhibitor of mitochondrial transport systems.
H Meisner et al.
Biochemistry, 13(14), 2795-2800 (1974-07-02)
Abdelhamid Abbas et al.
International journal of food microbiology, 161(3), 172-181 (2013-01-22)
The ochratoxin A (OTA) polyketide synthase otapks gene has been cloned from Penicillium verrucosum. A P. verrucosum mutant in which the otapksPV gene has been interrupted cannot synthesize ochratoxin A. The protein is most similar to the citrinin polyketide synthase
S C Duarte et al.
Food chemistry, 138(2-3), 1055-1061 (2013-02-16)
A reliable exposure assessment of mycotoxin contamination relies on their unambiguous identification and accurate quantification. With such purpose, a new analytical methodology was developed for evaluation of ochratoxin A (OTA) contamination in pork. Briefly OTA extraction from minced muscle samples
Xia Geng et al.
Analytical and bioanalytical chemistry, 405(8), 2443-2449 (2013-01-31)
By taking advantage of the intrinsic fluorescence of ochratoxin A (OTA), we present a fluorescence anisotropy approach for rapid analysis of the interactions between OTA and aptamers. The specific binding of OTA with a 36-mer aptamer can induce increased fluorescence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service