Skip to Content
Merck
All Photos(1)

Key Documents

62287

Sigma-Aldrich

Lipase A Candida antarctica, recombinant from Aspergillus oryzae

powder, beige, ~2 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.26

recombinant

expressed in Aspergillus oryzae

Quality Level

form

powder

specific activity

~2 U/mg

shelf life

limited shelf life, expiry date on the label

color

beige

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lipase A Candida antarctica (CALA) belongs to the α/β hydrolase family. The catalytic triad comprises Ser184, His366, and Asp334 residues.

Application

Lipase A Candida antarctica, recombinant from Aspergillus oryzae has been used in the enzyme-catalyzed hydrolysis studies with 3-acetylthiohexanal. It has also been used as a biocatalyst to test its effect on the esterification of phytosterol with caprylic acid.

Biochem/physiol Actions

Lipase A Candida antarctica (CALA) is a thermostable serine hydrolase with a higher sn-2 preference towards triglycerides. It displays excellent activity towards large substrates as well as bulky sterically hindered tertiary alcohols.
Lipase A from Candida Antarctica catalyzes hydrolysis of tertiary alcohols to form glycerol and fatty acids and shows selectivity for the N-acylation of β-amino esters.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol oleic acid per minute at pH 8.0 and 40°C (triolein, Cat. No. 62314, as substrate); 1 U as described above is equivalent to ~0.15 U using trybutyrine, Cat. No. 91010, as substrate, at pH 8.0 and 70°C

Other Notes

Characterization

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Biotechnological relevance of the lipase A from Candida antarctica
Monteiro RRC, et al.
Catalysis Today, 362, 141-154 (2021)
Anders G Sandström et al.
Protein engineering, design & selection : PEDS, 22(7), 413-420 (2009-06-11)
We herein report the first directed evolution of Candida antarctica lipase A (CalA), employing a combinatorial active-site saturation test (CAST). Wild-type CalA has a modest E-value of 5.1 in kinetic resolution of 4-nitrophenyl 2-methylheptanoate. Enzyme variants were expressed in Pichia
Integrated enzymatic production of specific structured lipid and phytosterol ester compositions
Hellner G, et al.
Process. Biochem., 45(8), 1245-1250 (2010)
S.A. Patkar et al.
Indian Journal of Chemistry, 76-76 (1993)
Hidehiko Wakabayashi et al.
Journal of agricultural and food chemistry, 51(15), 4349-4355 (2003-07-10)
The enantioselectivity of the generation of 3-mercaptohexanal and 3-mercaptohexanol, two potent sulfur-containing aroma compounds, by lipase-catalyzed hydrolysis of the corresponding 3-acetylthioesters was investigated. The stereochemical course of the kinetic resolutions was followed by capillary gas chromatography using modified cyclodextrins as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service