Skip to Content
Merck
All Photos(3)

Key Documents

306436

Sigma-Aldrich

Sudan Blue II

Dye content 98 %

Synonym(s):

Solvent Blue 35

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O2
CAS Number:
Molecular Weight:
350.45
Colour Index Number:
61554
Beilstein:
2398560
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder or crystals

Quality Level

composition

Dye content, 98%

technique(s)

microbe id | staining: suitable

mp

120-122 °C (lit.)

solubility

chloroform: 4 mg/mL, clear to opaque, blue

ε (extinction coefficient)

≥16000 at 648-652 nm in chloroform at 0.01 g/L

λmax

≥13000 at 600-604 nm in chloroform at 0.01 g/L (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCCCNc1ccc(NCCCC)c2C(=O)c3ccccc3C(=O)c12

InChI

1S/C22H26N2O2/c1-3-5-13-23-17-11-12-18(24-14-6-4-2)20-19(17)21(25)15-9-7-8-10-16(15)22(20)26/h7-12,23-24H,3-6,13-14H2,1-2H3

InChI key

OCQDPIXQTSYZJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sudan Blue II, also known as Solvent Blue 35 or Oil Blue 35, is a synthetic dye, with a functional azo group and aromatic ring. It is derived from anthraquinone.

Application

Sudan Blue II is used as a dye for staining alcohols, ester, hydrocarbon derivatives, oils, fats, and waxes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of Sudan Blue II in environmental samples after dispersive liquid-liquid microextraction
Unsal Y E
Quimica nova, 37 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service