Skip to Content
Merck
All Photos(1)

Key Documents

07169

Supelco

BTEX Standard

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

density

0.998 g/mL at 20 °C

application(s)

environmental

format

neat

storage temp.

2-8°C

Components

solution contains benzene, toluene, ethylbenzene, p-xylene, m-xylene, o-xylene, 10 μg/ml each in water

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service