Skip to Content
Merck
All Photos(3)

Key Documents

MABS482

Sigma-Aldrich

Anti-PL Scramblase 1 Antibody, clone 1A8

clone 1A8, from mouse

Synonym(s):

Phospholipid scramblase 1, PL Scramblase 1, Ca(2+)-dependent phospholipid scramblase 1, Erythrocyte phospholipid scramblase, MmTRA1b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
1A8, monoclonal
application:
IHC
IP
WB
species reactivity:
human, mouse
technique(s):
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
1

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A8, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PLSCR1(5359)

General description

Phospholipid scramblase 1 (UniProt O15162; also known as Ca(2+)-dependent phospholipid scramblase 1, Erythrocyte phospholipid scramblase, MmTRA1b, PL scramblase 1) is encoded by the PLSCR1 (also known as MMTRA1B) gene (Gene ID 5359) in human. Plasma membrane phospholipids are distributed asymmetrically between the inner and outer leaflets. Such asymmetrical distribution collapses in response to blood coagulation and apoptosis, resulting in phospholipid “scrambling” between the two leaflets of the plasma membrane. Flippases, floppases, and scramblases are three types enzymes known to mediate transbilayer lipid motion. Flippases and floppases function via ATP-dependent mechanism, while scramblases mediate transbilayer movement in a non-selective and energy-independent manner. Originally identified in 1996 as a 37 kDa erythrocyte type II transmembrane protein that mediates calcium-dependent membrane phospholipids redistribution, PL Scramblase 1 is the protein product encoded by the founding member of the PLSCR family of genes (PLSCR1-5). All PLSCR family members, with the exception of PLSCR2, possess a proline-rich N-terminal region containing PxxP and PPxY domains, a cysteine-rich region, a conserved calcium-binding domain (EF-hand-like), and a putative transmembrane region enriched in hydrophobic amino acids. In addition, PLSCR1 contains a nuclear localization signal (NLS) and a DNA-binding domain that are essential for its nuclear localization and associated nuclear function. In addition to PLSCRs, TMEM16 and XKR family members have also been reported to mediate scramblase activity.

Immunogen

Recombinant protein corresponding to human PL Scramblase 1.

Application

Anti-PL Scramblase 1 Antibody, clone 1A8 is an antibody against PL Scramblase 1 for use in Western Blotting, Immunohistochemistry, Immunoprecipitation.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected PL Scramblase 1 in 10 µg of A549 cell lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected PL Scramblase 1 in mouse colon tissue.
Western Blotting Analysis: A representative lot detected PL scramblase 1 in platelets and lung tissue from wild-type and PLSCR3-/-, but not PLSCR1-/-, mice, while little or no PL scramblase 1 was detected in adipocyte or muscle samples from wild-type or the knockout animals (Wiedmer, T., et al. (2004). Proc Natl Acad Sci USA. 101(36):13296-13301).
Western Blotting Analysis: A representative lot detected retroviral-mediated ectopic expression of murine PLSCR1 in SCF-ER-Hoxb8-immortalized myeloid progenitor cells from PLSCR1−/− mice (Chen, C.W., et al. (2011). J Leukoc Biol. 90(2):221-233).
Immunprecipitation Analysis: A representative lot immunoprecipitated PL scramblase 1 from murine bone marrow-derived mast cells (BMMCs) following IgE receptor activation. Tyrosine phosphorylation of PL scramblase 1 was then checked by Western blotting with clone 4G10 (Kassas, A., et al. (2014). PLoS One. 9(10):e109800).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected PL Scramblase 1 in 10 µg of HeLa cell lysate.

Target description

~35 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genomewide CRISPR knockout screen identified PLAC8 as an essential factor for SADS-CoVs infection.
Tse, et al.
Proceedings of the National Academy of Sciences of the USA, 119, e2118126119-e2118126119 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service