Skip to Content
Merck
All Photos(1)

Key Documents

MABN1837

Sigma-Aldrich

Anti-AGE Antibody, Carboxy-Methyl Lysine Antibody, clone 6C7

clone 6C7, from mouse

Synonym(s):

N(epsilon)-(carboxymethyl) lysine, CML, AGE-CML, N-epsilon-CML, AGE-CML (6C7) Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C7, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

rat, human

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG2aκ

shipped in

ambient

target post-translational modification

unmodified

General description

Chronic accumulation of Advanced Glycation End products (AGEs) is a common feature during aging, hyperlipidemia, and diabetes and affects function of proteins, lipids, and DNA. Although formation of AGE products is considered to be predominantly an endogenous process, certain external factors, such as some dietary products and smoking can increase the level of AGE in tissues. Under certain conditions glucose may form covalent adducts with the plasma proteins through the process of glycation. Glycation is a non-enzymatic reaction of reducing carbohydrates with lysine side chains and N-terminal amino groups of proteins. The term AGE often refers to non-reactive terminal products, such as N- carboxymethyllysine (CML) as well as reactive precursors, such as methylglyoxal (MG). CML is an AGE product that is usually found on proteins and lipids as a result of oxidative stress and chemical glycation. Excessive glycation can disrupt protein function by altering enzyme activity, receptor function, and molecular conformation. Protein glycation and subsequent formation of AGE products contributes to the pathogenesis of diabetes-related complications, such as retinopathy, nephropathy, and neuropathy. Higher levels of CML have been linked to greater hip fracture risk in older adults, independent of bone mineral density. AGE products interact with a variety of cell-surface AGE-binding receptors (RAGE) that results in their endocytosis and degradation. It can also generate pro-oxidant and pro-inflammatory events in cells. AGE products could also contribute to the development or worsening of many degenerative diseases, including Alzheimer′s disease. (Ref.: Cai, W., et al. (2002). Mol. Med 8 (7): 337-346; Barzilay, JI et al (2014). J. Bone Miner. Res. 29(5):1061-1066).

Specificity

Clone 6C7 specifically reacts with carboxymethyl lysine modified proteins.

Immunogen

Carboxymethyl Lysine ­modified Keyhole Limpet Hemocyanin (KLH).

Application

Anti-AGE, Carboxy-Methyl Lysine, clone 6C7, Cat. No. MABN1837, is a highly specific mouse monoclonal antibody, that targets Carboxy-Methyl Lysine (CML) modified proteins and has been tested in Immunohistochemistry (Paraffin).
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry in mouse kidney tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected AGE, Carboxy-Methyl Lysine in kidney tissue of mice fed high AGE diets.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J B J Kamphuis et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 36(5), e22297-e22297 (2022-04-09)
Irritable bowel syndrome (IBS) is a functional gastrointestinal disorder for which dietary interventions can be a useful treatment. In recent years, the low-FODMAP approach is gaining traction in this regard. The fermentation of these non-absorbed carbohydrates by the gut microbiota
Xi-Yuan Liu et al.
Frontiers in neuroscience, 15, 738022-738022 (2021-11-26)
Retinal pigment epithelium (RPE) serves critical functions in maintaining retinal homeostasis. An important function of RPE is to degrade the photoreceptor outer segment fragments daily to maintain photoreceptor function and longevity throughout life. An impairment of RPE functions such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service