Skip to Content
Merck
All Photos(1)

Key Documents

8.22098

Sigma-Aldrich

Methylstyrene

(mixture of 3- and 4-isomers) stabilised with 4-tert-butylpyrocatechol for synthesis

Synonym(s):

Methylstyrene, Vinyltoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3/4-(CH3)C6H4CH=CH2
CAS Number:
Molecular Weight:
118.18
MDL number:
UNSPSC Code:
12352005
EC Index Number:
215-292-7
NACRES:
NA.22

vapor pressure

1.47 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

575 °C

potency

5000 mg/kg LD50, oral (Rat)

expl. lim.

1.1-5.2 % (v/v)

mp

-77 °C

transition temp

flash point 52 °C

density

0.893 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

Cc1ccc(cc1)C=C

InChI

1S/C9H10/c1-3-9-6-4-8(2)5-7-9/h3-7H,1H2,2H3

InChI key

JLBJTVDPSNHSKJ-UHFFFAOYSA-N

Analysis Note

Assay 3-methyl styrol (GC, area%): 40.0 - 60.0 % (a/a)
Assay 4-methyl styrol (GC, area%): 40.0 - 60.0 % (a/a)
Sum isomers (GC): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.896 - 0.898
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

125.6 °F

Flash Point(C)

52 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service