Skip to Content
Merck
All Photos(1)

Key Documents

8.20576

Sigma-Aldrich

N-Fluoren-2-ylacetamide

for synthesis

Synonym(s):

N-Fluoren-2-ylacetamide, 2-Acetamidofluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-188-6
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

powder

mp

193-196 °C

storage temp.

2-30°C

SMILES string

N(c1cc2c(cc1)c3c(cccc3)C2)C(=O)C

InChI

1S/C15H13NO/c1-10(17)16-13-6-7-15-12(9-13)8-11-4-2-3-5-14(11)15/h2-7,9H,8H2,1H3,(H,16,17)

InChI key

CZIHNRWJTSTCEX-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 193 °C
Melting range (upper value): ≤ 196 °C
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service