Skip to Content
Merck
All Photos(1)

Key Documents

551476

Sigma-Aldrich

Q-VD-OPh, Non-O-methylated

≥90% (HPLC), liquid, Caspase inhibitor, Calbiochem®

Synonym(s):

InSolution Q-VD-OPh, Non-O-methylated, N-(2-Quinolyl)valyl-aspartyl-(2,6-difluorophenoxy)methyl Ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H25F2N3O6
Molecular Weight:
513.49
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Q-VD-OPh, Non-O-methylated, InSolution, ≥90%, irreversible broad-spectrum inhibitor of caspases

Quality Level

Assay

≥90% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable, irreversible, broad-spectrum caspase inhibitor (IC50 = 50, 100, 430, and <25 nM for caspase-1,-8, -9, and -3, respectively) with effective antiapoptotic properties against all major caspase-mediated cellular apoptosis pathways. Exhibits no cytotoxic effects even at extremely high concentrations. Shown to reduce ischemic brain damage and stroke-induced programmed cell death in thymus and spleen.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-1
Product does not compete with ATP.
Reversible: no
Target IC50: 50, 100, 430, and <25 nM for caspase-1,-8, -9, and -3, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Sequence

Q-Val-Asp-CH₂-OPh

Physical form

Supplied as a 10 mM (1 mg/195 µl) solution in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Braun, J.S., et al. 2007. Exp. Neurol.206, 183.
Caserta, T.M., et al. 2003. Apoptosis8, 345.
Rebbaa, A., et al. 2003. Oncogene22, 2805.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara H Small et al.
Science signaling, 14(714), eaba2611-eaba2611 (2021-12-22)
Cytokine production is a critical component of cell-extrinsic responses to DNA damage and cellular senescence. Here, we demonstrated that expression of the gene encoding interleukin-19 (IL-19) was enhanced by DNA damage through pathways mediated by c-Jun amino-terminal kinase (JNK) and
Javier Chicote et al.
Frontiers in pharmacology, 11, 580343-580343 (2020-11-13)
Macroautophagy (hereafter autophagy) is a multistep intracellular catabolic process with pleiotropic implications in cell fate. Attending to its activation, autophagy can be classified into inducible or constitutive. Constitutive, or basal autophagy, unfolds under nutrient-replete conditions to maintain the cellular homeostasis.
Yuanjiu Lei et al.
Cell, 186(14), 3013-3032 (2023-06-24)
Mitochondrial DNA (mtDNA) is a potent agonist of the innate immune system; however, the exact immunostimulatory features of mtDNA and the kinetics of detection by cytosolic nucleic acid sensors remain poorly defined. Here, we show that mitochondrial genome instability promotes
Joshua D Bryant et al.
Current protocols, 2(2), e372-e372 (2022-02-18)
Mitochondria have emerged as key drivers of mammalian innate immune responses, functioning as signaling hubs to trigger inflammation and orchestrating metabolic switches required for phagocyte activation. Mitochondria also contain damage-associated molecular patterns (DAMPs), molecules that share similarity with pathogen-associated molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service