Skip to Content
Merck
All Photos(2)

Key Documents

5.33917

Sigma-Aldrich

Choline Transporter Inhibitor II, ML352

Synonym(s):

Choline Transporter Inhibitor II, ML352, N-((3-Isopropylisoxazol-5-yl)methyl)-4-methoxy-3-((1-methylpiperidin-4-yl)oxy)benzamide, 4-Methoxy-3-(1-methylpiperidin-4-yl)oxy-N-((3-propan-2-yl-1,2-oxazol-5-yl)methyl)benzamide, CHT Inhibitor II, VU0476328

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H29N3O4
CAS Number:
Molecular Weight:
387.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable pyridinylpropanylthiophene-carboxamide compound that exhibits selective affinity toward MT/microtubule-bound KIFC1 motor domain (Q305-K673; Kd = 690 nM), but not to uncomplexed KIFC1 or MT, and selectively inhibits MT-induced, but not basal, KIFC1 motor domain ATPase activity (IC50 = 300 nM; [KIFC1] = 30 nM, [MT] = 100 nM, [ATP] = 15 µM) in an ATP-competitive (Ki = 43 nM; [KIFC1] = 125 nM, [MT] = 1.5 µM) and MT-noncompetitive manner, effectively preventing ADP release, while displaying little potency against 9 other kinesin motor proteins (<15% or no inhibition at 5 µM against human CENP-E, Chromokinesin/KIF4A, Eg5, KIFC3, KIF3C, KInesin Heavy Chain/KIF5B, MCAK/KIF2C, MKLP1/KIF23, and A. nidulans BimC/KIF8). KIFC1 knockdown by siRNA or functional inhibition by AZ82 (0.4 to 1.2 µM) is shown to result in mitotic delay and multipolar spindle formation among BT549 population with extra centrosomes due to failure of chromosomes clustering. In cells with normal/two centrosomes, Eg5 inhibition causes monopolar spindles formation, while AZ82 co-treatment (400 nM) offsets the effect of Eg5 inhibition (5 nM AZD4877) and restores bipolar spindles phenotype in mitotic HeLa cells. Off-target cytotoxicity is reported when used at concentrations above 4 µM.
A moderately brain permeant, non-choline based isoxazolylbenzamide compound that acts as a highly selective and potent inhibitor of high-affinity choline transporter (CHT; Ki = 92 nM in hCHT-LV-AA transfected HEK293 cells and 166 nM in mouse brain synaptosomes). The inhibition appears to be allosteric and noncompetitive. Does not affect the activity of acetylcholinesterase or cholineacetyltransferase and has no activity against dopamine, serotonin, and norepinephrine transporters. Also does not affect CHT protein expression. Exhibits high selectivity when tested against 68 GPCRs, ion channels, and transporters (IC50 >10 µM). Displays moderate pharmacokinetic properties (t1/2= 33 min, AUC = 77.5 ng/h/ml; Cmax = 32.6 ng/ml, at 2.3 mg/kg, p.o. in Sprague-Dawley rats) and attractive solubility (98.2 µM in PBS). Does not affect the activity of cytochrome P450 and hERG (IC50 >30 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CHT
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ennis, E.A., et al. ACS Chem. Neurosci.6, In press.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service