Skip to Content
Merck
All Photos(1)

Key Documents

5.30799

Sigma-Aldrich

AGE Inhibitor, LR-90

Synonym(s):

AGE Inhibitor, LR-90, Advanced Glycation Endproducts Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H34Cl2N4O8
CAS Number:
Molecular Weight:
709.57
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable methylene bisphenylureido derived compound that blocks the formation of advanced glycation endproduct (AGE) by scavenging dicarbonyl intermediates and by chelating transition metals that catalyze the production of AGE. Inhibits metal ion-catalyzed oxidation of ascorbate (IC50 = 275 µM). Exhibits protective effects against diabetic nephropathy, retinopathy, and atherosclerosis and reduces arterial stiffness in diabetic animals (~50 mg/L drinking water) by reducing collagen accumulation and cross-linking. Although it does not affect glycemic condition and body weight in normal and diabetic animals, it reduces plasma triglycerides and glycated hemoglobin levels in diabetic rats and improves their cardiac output and stroke volume. Also shown to suppress S100b and TNFa-induced activation of NF-κB.

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A cell-permeable methylene bisphenylureido derived compound that blocks the formation of advanced glycation endproduct (AGE) by scavenging dicarbonyl intermediates and by chelating transition metals that catalyze the production of AGE. Inhibits metal ion-catalyzed oxidation of ascorbate (IC50 = 275 µM). Exhibits protective effects against diabetic nephropathy, retinopathy, and atherosclerosis and reduces arterial stiffness in diabetic animals (~50 mg/L drinking water) by reducing collagen accumulation and cross-linking. Although it does not affect glycemic condition and body weight in normal and diabetic animals, it reduces plasma triglycerides and glycated hemoglobin levels in diabetic rats and improves their cardiac output and stroke volume. Also shown to suppress S100b and TNFa-induced activation of NF-kB.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
=IF(SGM_MetricTransfer!C11=,,SGM_MetricTransfer!C11)

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Satheesan, S., et al. 2014. Br. J. Pharmacol.171, 3103.

Nagai, R., et al. 2012. Diabetes.61, 549.
Figarola, J.L., et al. 2008. Diabetologia.51, 882.
Figarola, J.L., et al. 2003. Diabetologia.46, 1140.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service