5.00440
Refractive index standard
traceable to NIST, traceable to PTB, [α]/D 40°, Brix CERTIPUR®
About This Item
Recommended Products
grade
analytical standard
Quality Level
Agency
traceable to NIST
traceable to PTB
product line
Certipur®
optical activity
[α]/D 40°
manufacturer/tradename
Brix CERTIPUR®
application(s)
environmental
flavors and fragrances
food and beverages
general analytical
storage temp.
15-25°C
General description
Application
- Refractive index standard for use as a reference standard in determination of Brix value by refractometers
- The use of this standard for determining sugar content is also possible in the wine and beer, sugar, fruit juice, honey, starch, and sugar manufacturing industries
Features and Benefits
- Each product has five ampoules, and each ampoule contains an 8ml solution
- Accompanied by technical data sheet that includes information about extended measurement uncertainty calculated according to ISO 17025
- Traceable to recognized national standards, and to units of measurement recognized at the National Institute Standards and Technology (NIST) and Physical Technical Institute (PTB)
- Solutions that are stable, ready to use, and have long minimum shelf lives
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service