Skip to Content
Merck
All Photos(2)

Key Documents

444295

Sigma-Aldrich

MT1-MMP Inhibitor, NSC405020

The MT1-MMP Inhibitor, NSC405020 controls the biological activity of MT1-MMP. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MT1-MMP Inhibitor, NSC405020, 3,4-Dichloro-N-(pentan-2-yl)benzamide, 3,4-Dichloro-N-(1-methylbutyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15Cl2NO
CAS Number:
Molecular Weight:
260.16
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C=C(C(NC(C)CCC)=O)C=C1

InChI key

ARDYECYBETXQFD-UHFFFAOYSA-N

General description

A cell-permeable pentanylbenzamide compound that acts as an allosteric, reversible, and selective inhibitor of the collgenolytic activity of MT1-MMP. Does not affect the catalytic activity of cellular MT-1-MMP thus allows it to retain its ability to activate MMP-2. Selectively targets the hemopexin (PEX) domain of MT1-MMP, thereby repressing its pro-tumorigenic activity. Its binding to the PEX domain is dependent on Met-328, Arg-330, Asp-376, Met-422, and Ser-470 in the druggable pocket of the enzyme. Shown to block 184B5-MT cell migration on collagen-1 and repress MCF7-β3/MT tumor growth in xenografted mice (0.5 mg/kg, intratumoral).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Other Notes

Remacle, A.G., et al. 2012. Cancer Res.72, 2339.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrés Alberro-Brage et al.
Frontiers in immunology, 14, 1260973-1260973 (2023-09-20)
Acute respiratory distress syndrome (ARDS) is a common complication of influenza virus (IV) infection. During ARDS, alveolar protein concentrations often reach 40-90% of plasma levels, causing severe impairment of gas exchange and promoting deleterious alveolar remodeling. Protein clearance from the
Cyril Andrieu et al.
Development (Cambridge, England), 147(7) (2020-04-14)
Matrix metalloproteinases have a broad spectrum of substrates ranging from extracellular matrix components and adhesion molecules to chemokines and growth factors. Despite being mostly secreted, MMPs have been detected in the cytosol, the mitochondria or the nucleus. Although most of

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service