Skip to Content
Merck
All Photos(1)

Key Documents

386040

Sigma-Aldrich

Anti-Hsp90 Mouse mAb (AC88)

liquid, clone AC88, Calbiochem®

Synonym(s):

Anti-Hsp82, Anti-Hsp85, Anti-Hsp89

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
clone:
AC88, monoclonal
application:
citations:
4

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

AC88, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity (predicted by homology)

all

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSP90AA1(3320)

General description

Protein G purified mouse monoclonal antibody. Recognizes the ~90 kDa Hsp90 protein.
Recognizes the ~90 kDa Hsp90 protein.
This Anti-Hsp90 Mouse mAb (AC88) is validated for use in Immunoblotting, Immunoprecipitation, Immunocytochemistry for the detection of Hsp90.

Immunogen

Achyla ambisexualis
Hsp90 isolated from water mold, Achyla ambisexualis

Application

Immunoblotting (1 µg/ml, colorimetric)

Immunoprecipitation (see comments)

Immunocytochemistry (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 50% glycerol, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Heat shocked HeLa cells, Heat shocked 3T3 cells, Heat shocked PC-12 cells, Heat shocked CHO-K1, Heat shocked Rat-2 cells

Other Notes

Cross-reacts with glucocorticoid receptor in rodent. By immunoprecipitation, the antibody favors free Hsp90 over complexed Hsp90. The optimal dilution for immunoprecipitation is assay dependent and must be determined for each sample. This antibody has also been reported to work for immunocytochemistry. Variables associated with assay conditions will dictate the optimal dilution.
Harry, J.L. et al. 1990. Development190, 305.
Minoo, P. et al. 1989 J. Cell. Biol.109, 1937.
Riehl, R.M., et al. 1985. Biochemistry 24, 6586.
Sanchez, E.R., et al. 1985. J. Biol. Chem. 260, 12398.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Shaikho et al.
Translation (Austin, Tex.), 4(2), e1244395-e1244395 (2017-01-17)
Mammalian 90 kDa heat shock protein (Hsp90) is a ubiquitous molecular chaperone whose expression is selectively upregulated during stress, although the precise control mechanism of this increase is yet to be fully elucidated. We used polysome profiling to show that Hsp90α
Bahram Samanfar et al.
PeerJ, 5, e4037-e4037 (2017-11-22)
The presence of acetic acid during industrial alcohol fermentation reduces the yield of fermentation by imposing additional stress on the yeast cells. The biology of cellular responses to stress has been a subject of vigorous investigations. Although much has been
Tania Martins-Marques et al.
Life science alliance, 3(12) (2020-10-25)
Ischemic heart disease has been associated with an impairment on intercellular communication mediated by both gap junctions and extracellular vesicles. We have previously shown that connexin 43 (Cx43), the main ventricular gap junction protein, assembles into channels at the extracellular
Kushaan Dumasia et al.
Epigenetics, 12(11), 953-963 (2017-09-28)
Estrogen receptors (ESR1 and ESR2) play crucial roles in various processes during spermatogenesis. To elucidate individual roles of ESRs in male fertility, we developed in vivo selective ESR agonist administration models. Adult male rats treated with ESR1 and ESR2 agonist

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service