Skip to Content
Merck
All Photos(1)

Key Documents

341610

Sigma-Aldrich

FGF/PDGF/VEGF RTK Inhibitor

The FGF/PDGF/VEGF RTK Inhibitor, also referenced under CAS 144335-37-5, controls the biological activity of FGF/PDGF/VEGF RTK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

FGF/PDGF/VEGF RTK Inhibitor, 4-Hydroxy-3-benzimidazol-2-ylhydroquinolin-2-one, PDGFR Tyrosine Kinase Inhibitor XVII, VEGFR Tyrosine Kinase Inhibitor XXX, VEGFR2 Kinase Inhibitor XXVIII, PDGFR Tyrosine Kinase Inhibitor XVII, 4-Hydroxy-3-benzimidazol-2-ylhydroquinolin-2-one, VEGFR Tyrosine Kinase Inhibitor XXX, VEGFR2 Kinase Inhibitor XXVIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N3O2
CAS Number:
Molecular Weight:
277.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1c2c(nc1c3c(nc4c(c3O)cccc4)O)cccc2

InChI

1S/C16H11N3O2/c20-14-9-5-1-2-6-10(9)19-16(21)13(14)15-17-11-7-3-4-8-12(11)18-15/h1-8H,(H,17,18)(H2,19,20,21)

InChI key

YXCWBNCDJZEBKY-UHFFFAOYSA-N

General description

A cell-permeable benzimidazolo-hydroquinolinone compound that acts as a potent, reversible, ATP-competitive inhibitor against PDGFRβ, FGFR-1, and VEGFR-2 (IC50 = 20, 90, and 240 nM, respectively) and effectively suppresses VEGF-stimulated proliferation of HMVECs (EC50 = 420 nM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Renhowe, P.A., et al. 2009. J. Med. Chem.52, 278.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service