Skip to Content
Merck
All Photos(1)

Key Documents

324402

Sigma-Aldrich

Anti-Dopamine D₃ Receptor Rabbit pAb

lyophilized, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
clone:
polyclonal
application:
species reactivity:
rat, human
citations:
2

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

does not contain

preservative

species reactivity

rat, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... DRD3(1814)
rat ... Drd3(29238)

General description

Anti-Dopamine D₃ Receptor, rabbit polyclonal, recognizes the dopamine D3 receptor in rat brain. Does not cross-react with other dopamine receptors. It is validated for use in ELISA, WB, FC, and IHC.
Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the ~50 kDa D3 dopamine receptor protein.
Recognizes the dopamine D3 receptor in rat brain. Does not cross-react with other dopamine receptors.

Immunogen

a synthetic peptide (ASLSQLSSHC) corresponding to amino acids (2-10) of human dopamine D₃ receptor

Application

ELISA (see comments)

Flow Cytometry (1:100, see application references)

Immunoblotting (1:500)

Immunohistochemistry (1:5000)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Reconstitute the antibody with 100 µl PBS containing 10 mg/ml BSA. For further dilution of the antibody use a similar buffer formulation. Following reconstitution, refrigerate (4°C).

Other Notes

This antiserum is highly specific for the immunizing peptide sequence. For immunohistochemistry, a 1:5000 dilution of the reconstituted serum demonstrated staining of specific cells in various regions of rat brain sections. Such staining includes dendritic labeling in the molecular layers (cortex layer 1 and molecular layer of the hippocampus) as well as the plasma membranes of neurons in layer 5 of cortex and the glomeruli of the olfactory bulb. This antibody has also been reported to work for ELISA. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacqueline S Coley et al.
PloS one, 10(2), e0117450-e0117450 (2015-02-04)
Drug abuse is a major comorbidity of HIV infection and cognitive disorders are often more severe in the drug abusing HIV infected population. CD14+CD16+ monocytes, a mature subpopulation of peripheral blood monocytes, are key mediators of HIV neuropathogenesis. Infected CD14+CD16+
Tina M Calderon et al.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology, 12(2), 353-370 (2017-01-31)
In human immunodeficiency virus-1 (HIV) infected individuals, substance abuse may accelerate the development and/or increase the severity of HIV associated neurocognitive disorders (HAND). It is proposed that CD14+CD16+ monocytes mediate HIV entry into the central nervous system (CNS) and that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service