Skip to Content
Merck
All Photos(1)

Key Documents

181198

Sigma-Aldrich

Arachidonic Acid

≥99% (TLC and GC), oil, membrane phospholipid component, Calbiochem®

Synonym(s):

Arachidonic Acid, cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic Acid, 20:4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O2
CAS Number:
Molecular Weight:
304.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Arachidonic Acid, Precursor for prostaglandins, prostacyclin, and thromboxane.

description

Merck USA index - 14, 765

Quality Level

Assay

≥99% (TLC and GC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

colorless

solubility

ethanol: 5 mg/mL

shipped in

dry ice

storage temp.

−70°C

SMILES string

OC(=O)CCC\C=C\C\C=C\C\C=C\C\C=C\CCCCC

InChI

1S/C20H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-19H2,1H3,(H,21,22)/b7-6+,10-9+,13-12+,16-15+

InChI key

YZXBAPSDXZZRGB-CGRWFSSPSA-N

General description

A cis-polyunsaturated fatty acid found in the plasma membrane. Major component of membrane phospholipids. Following its release by cellular phospholipases, it is enzymatically converted to a variety of inflammatory and vasoactive metabolites, such as prostaglandins (via the cyclooxygenase pathway), leukotrienes (via the lipoxygenase pathway), and epoxides (via the cytochrome P450 pathway). Binds to G-protein α-subunits in a covalent, posttranslational manner. Inhibits Ras-GAP. Stimulates the synthesis of nitric oxide by platelets.
Precursor for prostaglandins, prostacyclin, and thromboxane. Binds to G-protein α-subunits in a covalent, post-translational manner. Inhibits Ras-GAP. Stimulates the synthesis of nitric oxide by platelets. Modulates nitric oxide production by prostaglandin synthesis via the cyclooxygenase pathway.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Ras-GAP
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Hallak, H., et al. 1994. J. Biol. Chem. 269, 4713.
Khurana, G., and Bennett, M.R. 1993. Br. J. Pharmacol. 109, 480.
Han, J.-W., et al. 1991. Science252, 576.
Radomski, M.W., et al. 1990. Br. J. Pharmacol. 101, 325.
Neddleman P., et al. 1986. Annu. Rev. Biochem.55, 69.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. What is the preferred method for diluting and aliquoting the compound?

    1 answer
    1. This product is soluble in ethanol at a concentration of 5 mg/mL. After reconstitution, aliquot and freeze at -20°C. Stock solutions remain stable for up to 1 month at -20°C.

      Helpful?

  2. What is the volume in uL of the 100mg AA vial? Why is it not stated on the vial?

    1 answer
    1. Please note that the density of this product is about 0.92 g/mL. Hence, 100 mg material has a volume of 108 microliters.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service