Skip to Content
Merck
All Photos(2)

Key Documents

16-318

Sigma-Aldrich

Anti-Akt/PKB Antibody, PH Domain, clone SKB1, Magnetic Bead Conjugate

clone SKB1, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
SKB1, monoclonal
application:
IP
species reactivity:
mouse, rat, human
technique(s):
immunoprecipitation (IP): suitable
citations:

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

SKB1, monoclonal

species reactivity

mouse, rat, human

technique(s)

immunoprecipitation (IP): suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Akt1(11651)

General description

Akt (protein kinase B), a serine/threonine kinase, has emerged as a critical enzyme in signal transduction pathways involved in cell proliferation, apoptosis, angiogenesis, and diabetes. In mammals three isoforms of Akt (α, β, γ or Akt 1, 2, 3) are reported that exhibit a high degree of homology, but differ slightly in the localization of their regulatory phosphorylation sites. Akt α is the predominant isoform in most tissues, whereas the highest expression of Akt β is observed in the insulin-responsive tissues, and Akt γ is abundant in brain tissue. Each Akt isoform is composed of three functionally distinct regions: an N-terminal pleckstrin homology (PH) domain that provides a lipid-binding module to direct Akt to PIP2 and PIP3, a central catalytic domain, and a C-terminal hydrophobic motif.

Specificity

Recognizes Akt1/PKBα.

Immunogen

GST-fusion protein corresponding to residues 1-149 of human Akt1

Application

Detect Akt/PKB using this Anti-Akt/PKB Antibody, PH Domain, clone SKB1, Magnetic Bead Conjugate validated for use in IP.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Quality

Evaluated by immunoprecipitation on 250 μg of NIH3T3 whole cell lysate stimulated with 50 ng/mL PDGF (5 minutes).

Target description

~ 60 kDa observed

Physical form

20% slurry; 1X PBS, 0.05% sodium azide.
Sufficient for 10 IPs.
Format: Purified
Protein G Chromatography

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
NIH3T3 whole cell lysate stimulated with 50 ng/mL PDGF (5 minutes)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service