Skip to Content
Merck
All Photos(2)

Key Documents

1.46284

Millipore

Tryptic Soy Broth- Ready-to-use Bags, Tubes & Ampoules

1/2 ×, tube capacity × filling volume 12 mL × 9 mL, closure type, blue screw cap with 2 loci, box of 100 tubes

Synonym(s):

CASO, Casein Soybean Digest Broth, SCDB, TSB, TSB 1/2 concentration, Tryptic Soy Broth half-concentrated, Tryptone Soy Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

sterility

sterile; autoclaved

Quality Level

form

solution

feature

closure type blue screw cap with 2 loci
ready-to-use

composition

, 1/2 ×

packaging

box of 100 tubes
tube of

capacity

12 mL

tube capacity × filling volume

12 mL × 9 mL

application(s)

pharmaceutical
sterility testing

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service