1.06756
Methyl 4-hydroxybenzoate sodium salt
EMPROVE® ESSENTIAL, Ph. Eur., BP, NF
Pharma Manufacturing
Synonym(s):
Methyl 4-hydroxybenzoate sodium salt, 4-Hydroxybenzoic acid methyl ester sodium salt, Methylparaben sodium salt
About This Item
Recommended Products
Agency
BP
NF
Ph. Eur.
Quality Level
product line
EMPROVE® ESSENTIAL
form
solid
autoignition temp.
>600 °C
pH
9.7-10.3 (1 g/L in H2O)
mp
>125 °C
density
1.42 g/cm3 at 20 °C
application(s)
liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation
storage temp.
15-25°C
SMILES string
[Na+].[O-]c1ccc(cc1)C(=O)OC
InChI
1S/C8H8O3.Na/c1-11-8(10)6-2-4-7(9)5-3-6;/h2-5,9H,1H3;/q;+1/p-1
InChI key
PESXGULMKCKJCC-UHFFFAOYSA-M
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
Disclaimer
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.
Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.
Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.
Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service