Skip to Content
Merck
All Photos(3)

Key Documents

04-963

Sigma-Aldrich

Anti-CTLA4 (CD152) Antibody, clone 9H10

clone 9H10, from hamster

Synonym(s):

CD152 antigen, Cytotoxic T-lymphocyte-associated antigen 4, celiac disease 3, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
9H10, monoclonal
application:
FACS
IP
WB
species reactivity:
mouse
technique(s):
flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
1

biological source

hamster

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9H10, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Related Categories

General description

CTLA4, Cytotoxic T lymphocyte antigen 4, also known as CD152, belongs to the family of immunoglobins. It is widely expressed on activated T lymphocytes and is involved in the maintenance of immune tolerance. CTLA4 is a homolog for CD28. They are able to bind to B7-1 and B7-2; however, CTLA4 is able to bind to both with a higher affinity. Defects in CTLA4 have been linked to Rheumatoid arthritis, Addisons disease, Graves disease and many other autoimmune diseases.

Specificity

This antibody recognizes CTLA4. Antibody 9H10 is specific for CTLA-4 by four criteria: (a) Reactivity against CTLA-4 Ig but not CD4 lg; (b) the ability to block CTLA-4 Ig binding to B7 transfectants; (c) the ability to stain activated T cells but not freshly isolated T cells; and (d) the ability to stain a CTLA-4 transfectant but not control transfectants (Krummel, M.F., et al., 1995).

Immunogen

Mouse CTLA-4/human IgG1 fusion protein corresponding to residues surrounding CTLA4.

Application

Anti-CTLA4 (CD152) Antibody, clone 9H10 is an antibody against CTLA4 (CD152) for use in WB, FC & IP.
Flow Cytometry Analysis: Representative images from a previous lot of primary antibody that was biotinylated. Results with 04-963 are expected to be similar if used directly with an anti-Syrian hamster conjugated antibody, however because of the low expression of CTLA4 Fab fragmented goat anti-Syrian hamster biotin followed SA-fluorophore is recommended for best results.
FACS analysis of ConA (24 hours) stimulated BALB/C splenocytes for surface expression of CTLA-4 (CD152). Cells were stained with 10 ug of biotin-labeled hamster anti-CTLA-4 ,clone 9H10 (open histogram) or with PE-Sav secondary antibody alone (histogram, Figure B). Note the distinct signal difference between the control (dark control histogram) and test (clear histogram).
Immunoprecipitation: Clone 9H10 is widely used for immunoprecipitaiton of CTLA-4; recommended lysis buffers include 1% NP40; 0.5% triton X-100 with proteinase inhibitors; 1-5 µg of 9H10 is added to 300-500 µL of whole cell lysate or cell membrane protein preparations (250-500 mg/mL totall protein) and incubated 3-6 hours at 4°C, followed by capture with protein G beads following standard immunoprecipitation methods (Zhang, Y., et al., 1997).
Research Category
Inflammation & Immunology
Research Sub Category
Transcription Factors

Inflammation & Autoimmune Mechanisms

Quality

Evaluated by Western Blot in mouse spleen lysate.
Western Blot Analysis: 1 µg/mL of a biotinylated version of this antibody detected CTLA4 in 10 µg of mouse spleen lysate.
THIS ANTIBODY IS NO BIOTINYLATED.

Target description

~30 kDa

Physical form

Format: Purified
Protein G Purified
Purified hamster monoclonal IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4) with 150 mM NaCl and 0.05% sodium azide.

Storage and Stability

Stable at 2-8°C for 1 year from date of receipt.

Analysis Note

Control
Mouse spleen cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Lagunas-Gomez et al.
Journal of cell science, 136(3) (2023-01-19)
The endoplasmic reticulum (ER) is the start site of the secretory pathway, where newly synthesized secreted and membrane proteins are packaged into COPII vesicles through direct interaction with the COPII coat or aided by specific cargo receptors. Little is known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service