Skip to Content
Merck
All Photos(1)

Key Documents

900412E

Avanti

oleic acid(17-yne)

Avanti Research - A Croda Brand 900412E, ethanol solution

Synonym(s):

(Z)-octadec-9-en-17-ynoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H30 O2
CAS Number:
Molecular Weight:
278.43
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

ethanol solution

packaging

pkg of 1 × 1 mL (900412E-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900412E

concentration

1 mg/mL (900412E-1mg)

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

General description

Oleic acid (17-yne) is a click chemistry reagent useful for membrane research. The clickable alkyne group may be used to probe cholesterol binding interactions in living cells.

Packaging

5 mL Clear Glass Sealed Ampule (900412E-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service