Skip to Content
Merck
All Photos(1)

Key Documents

810242C

Avanti

TopFluor TMR PS

Avanti Research - A Croda Brand 810242C

Synonym(s):

1-oleoyl-2-(6-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3α,4α-diaza-s-indacene-2-propionyl)amino)hexanoyl)-sn-glycero-3- phospho-L-serine (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H79 BF2N5 O12P
CAS Number:
Molecular Weight:
1033.98
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810242C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810242C

concentration

1 mg/mL (810242C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphoserine is a phosphoamino acid that is most abundant in the eukaryotic phosphoproteome. Phosphoserine is synthesized post-translationally as it is not encoded in the genetic code.

Biochem/physiol Actions

Phosphoserine (PS) is a metabolic precursor for serine. In some methanogenic archaea, PS is utilised to incorporate cysteine into proteins.

Packaging

5 mL Amber Glass Screw Cap Vial (810242C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel T Rogerson et al.
Nature chemical biology, 11(7), 496-503 (2015-06-02)
Serine phosphorylation is a key post-translational modification that regulates diverse biological processes. Powerful analytical methods have identified thousands of phosphorylation sites, but many of their functions remain to be deciphered. A key to understanding the function of protein phosphorylation is
Hee-Sung Park et al.
Science (New York, N.Y.), 333(6046), 1151-1154 (2011-08-27)
O-Phosphoserine (Sep), the most abundant phosphoamino acid in the eukaryotic phosphoproteome, is not encoded in the genetic code, but synthesized posttranslationally. Here, we present an engineered system for specific cotranslational Sep incorporation (directed by UAG) into any desired position in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service