Skip to Content
Merck
All Photos(2)

Key Documents

810141P

Avanti

18:0 NBD PE (NBD-DSPE)

Avanti Research - A Croda Brand

Synonym(s):

NBD-DSPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H86N5O11P
CAS Number:
Molecular Weight:
928.19
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810141P-1mg)
pkg of 5 × 2 mg (810141P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

18:0 NBD PE (NBD-DSPE) is a light sensitive lipid which is not hygroscopic. It has saturated acyl chains.
NBD-DSPE preferentially partitions, although not entirely, into the more ordered cholesterol and sphingolipid-enriched microdomain known as a lipid "raft".

Application

18:0 NBD PE (NBD-DSPE) has been used as NBD-labeled lipids to study the partitioning behavior in vesicles.

Packaging

5 mL Amber Glass Screw Cap Vial (810141P-10mg)
5 mL Amber Glass Screw Cap Vial (810141P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ordered raft domains induced by outer leaflet sphingomyelin in cholesterol-rich asymmetric vesicles
Lin Q and London E
Biophysical Journal (2015)
Ordered raft domains induced by outer leaflet sphingomyelin in cholesterol-rich asymmetric vesicles
Lin Q and London E
Biophysical Journal (2015)
Ordered raft domains induced by outer leaflet sphingomyelin in cholesterol-rich asymmetric vesicles
Lin Q and London E
Biophysical Journal, 2212-2222 (2015)
Peter J Quinn
Progress in lipid research, 49(4), 390-406 (2010-05-19)
Domains in cell membranes are created by lipid-lipid interactions and are referred to as membrane rafts. Reliable isolation methods have been developed which have shown that rafts from the same membranes have different proteins and can be sub-fractionated by immunoaffinity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service