Key Documents
Z680567
Caps for Duran® laboratory bottles
neck thread for GL 14, red PBT cap (with lip seal)
Synonym(s):
caps bottles, glass bottles caps
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
red PBT cap (with lip seal)
manufacturer/tradename
DURAN®
Schott
fitting
neck thread for GL 14
General description
The GL 14 caps and tubing inserts allow the DURAN® GL 45 2/3 port tubing connection caps or GL 45 / GLS 80® Stirred Bottle Reactors to be used with four different rigid or semi-rigid tubing outer diameters (1.6, 3.0, 3.2, or 6.0 mm). The different sized inserts form a tight seal around the pass-through tubing when used with the blue GL 14 tubing cap. Sterile pressure equalisation is possible through use of 0.2 µm syringe filter set. Unused ports can be sealed off with a red PBT GL 14 screw cap.
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Features and Benefits
• Manufactured from PP, PTFE, silicone• Accessories designed for use with the DURAN® GL 45 2/3 port tubing connection caps or GL 45 / GLS 80® Stirred Bottle Reactor caps• Safe and secure liquid transfer into or out of laboratory bottles• Four inserts are suitable for rigid or semi-rigid tubing with a range of outer diameters: 1.6, 3.0, 3.2 or 6.0 mm• Open-topped blue PP GL 14 pass-through tubing cap for use with the inserts• Closed-top red PBT GL 14 cap for sealing unused ports• Sterile pressure equilisation is possible with pressure compensation set
Legal Information
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)