Z677914
Finntip® Flex pipette tips
Finntip Flex 1000, non-sterile
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
maximum volume:
1 mL
volume range:
100-1000 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 1000 ea (Bag)
manufacturer/tradename:
Finn 94060720
Recommended Products
material
blue
colorless tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 1000 ea (Bag)
manufacturer/tradename
Finn 94060720
maximum volume
1 mL
volume range
100-1000 μL
Related Categories
General description
- Finntip® Flex pipette tips are compatible with both single and multichannel pipettes.
- Tips are DNA, DNase, RNase, and endotoxin-free.
- Offers consistent seal for better precision and accurate results.
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
Other Notes
Browse all pipette tips
Legal Information
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)