Z567485
NMR tube caps
NMR tube size 4 mm, blue
Synonym(s):
cap tubes, nmr tubes, plastic end cap
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
SB.32
Recommended Products
material
polypropylene
packaging
pkg of 100 ea
manufacturer/tradename
Wilmad 521-G-100
NMR tube size
4 mm
color
blue
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Disposable NMR Tube Caps are manufactured from molded PVC or Polyethylene depending on the tube OD. The different colors help track samples based on various reference solvents. When using Chloroform or Acetone as reference solution, avoid (1) direct contact between the sample and the disposable cap, (2) long term storage over 2 hours. If these conditions are inevitable, use PTFE NMR tube caps.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service