Z566136
Pigment-free, filler-free sleeve stopper septa
bottom I.D. × O.D. 4.2 mm × 10.7 mm
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
Recommended Products
material
amber septa
natural rubber septa
Joint
joint ST/NS10/30
feature
Use with small tubes
bottom I.D. × O.D.
4.2 mm × 10.7 mm
Related Categories
General description
These sleeve stopper septa are manufactured from pure natural rubber without fillers or pigments for super tactility and resealability. Less additives in the rubber lowers the potential for contamination during use.
Storage Note for Natural Rubber Septa: Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.
Storage Note for Natural Rubber Septa: Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.
Choose from one of the most recent versions:
Certificates of Analysis (COA)