Z177458
Scienceware® polypropylene scoops
capacity 200 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
Bel-Art F367530000
bowl size
127 mm × 51 mm × 51 mm
capacity
200 mL
handle size
65 mm
Looking for similar products? Visit Product Comparison Guide
General description
Molded, natural color.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service