Key Documents
WHA10534320
Whatman® protein saver cards
903 Protein saver snap apart card, pkg of 100 ea
Synonym(s):
Whatman fta, Z694827, dbs cards, pcr blood
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 10534320
Whatman Article No. 28416622 (US reference)
General description
The paper in our 903 Proteinsaver Snap-Apart Card, imprinted with four half-inch circles, is enclosed between two pieces of cover stock. Each circle holds 75-80 µl of sample. To use the 903 Proteinsaver Snap-Apart Card, one cover is snapped off, the sample is collected and the remaining cover is folded over the sample. This cover is imprinted with the universal biohazard symbol.
Features and Benefits
  • Ready-to-use cards and kits save time and costs during sample collection, transport, and archiving.
  • Strict quality control and ISO9001 manufacturing standards ensure high quality, reproducibility, and purity.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Customers Also Viewed
Samira Asgari et al.
Nature, 582(7811), 234-239 (2020-06-06)
On average, Peruvian individuals are among the shortest in the world1. Here we show that Native American ancestry is associated with reduced height in an ethnically diverse group of Peruvian individuals, and identify a population-specific, missense variant in the FBN1
Protocols
A number of options are available for sample collection, transport, archiving, and DNA purification.
A number of options are available for sample collection, transport, archiving, and DNA purification.
A number of options are available for sample collection, transport, archiving, and DNA purification.
A number of options are available for sample collection, transport, archiving, and DNA purification.