Skip to Content
Merck
All Photos(1)

Key Documents

W332402

Sigma-Aldrich

3,5,5-Trimethyl-1-hexanol

≥85%

Synonym(s):

3,5,5-trimethyl hexanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
144.25
FEMA Number:
3324
EC Number:
Council of Europe no.:
702
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.055
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥85%

refractive index

n20/D 1.432 (lit.)

bp

193-194 °C (lit.)

density

0.824 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; floral; herbaceous

SMILES string

CC(CCO)CC(C)(C)C

InChI

1S/C9H20O/c1-8(5-6-10)7-9(2,3)4/h8,10H,5-7H2,1-4H3

InChI key

BODRLKRKPXBDBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,5,5-Trimethyl-1-hexanol is a branched chain saturated alcohol that is commonly used as a fragrance ingredient. It occurs naturally in Lycii fructus.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aroma?Active Components of Lycii fructus (kukija).
Lee GH, et al.
Journal of Food Science, 73(6) (2008)
Fragrance material review on 3, 5, 5-trimethyl-1-hexanol.
McGinty D, et al.
Food And Chemical Toxicology, 48,, S47-S50 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service