Skip to Content
Merck
All Photos(1)

Key Documents

W288306

Sigma-Aldrich

1-Phenyl-3-methyl-3-pentanol

≥98%

Synonym(s):

lilac pentanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2883
EC Number:
Council of Europe no.:
86
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.037
NACRES:
NA.21
Organoleptic:
floral
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Agency

meets purity specifications of JECFA

Assay

≥98%

refractive index

n20/D 1.511

bp

112-112.5 °C/0.8 mmHg

density

0.938 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral

SMILES string

OC(CCc1ccccc1)(CC)C

InChI

1S/C12H18O/c1-3-12(2,13)10-9-11-7-5-4-6-8-11/h4-8,13H,3,9-10H2,1-2H3

InChI key

AEJRTNBCFUOSEM-UHFFFAOYSA-N

Related Categories

General description

1-Phenyl-3-methyl-3-pentanol is a fragrance component commonly used in cosmetics, toiletries, shampoos, fine fragrances, and non-cosmetic/household products like detergents and cleaners.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragrance material review on 1-phenyl-3-methyl-3-pentanol
Scognamiglio J, et al.
Food And Chemical Toxicology, 50, S168-S172 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service