W277704
Neryl propionate
≥98%, mixture of isomers, natural (US), FG
Synonym(s):
[(2Z)-3,7-dimethylocta-2,6-dienyl] propanoate
About This Item
Recommended Products
grade
FG
Halal
Kosher
natural (US)
Quality Level
Agency
follows IFRA guidelines
meets purity specifications of JECFA
reg. compliance
EU 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
Assay
≥98%
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
greener alternative category
Organoleptic
green; fruity; floral; sweet
SMILES string
O(C\C=C(/CCC=C(C)C)\C)C(=O)CC
InChI
1S/C13H22O2/c1-5-13(14)15-10-9-12(4)8-6-7-11(2)3/h7,9H,5-6,8,10H2,1-4H3/b12-9-
InChI key
BYCHQEILESTMQU-XFXZXTDPSA-N
General description
Other Notes
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service