Skip to Content
Merck
All Photos(1)

Key Documents

W275301

Sigma-Aldrich

Methyl isovalerate

≥99%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COOCH3
CAS Number:
Molecular Weight:
116.16
FEMA Number:
2753
Beilstein:
1699922
EC Number:
Council of Europe no.:
457
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.462
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥99%

refractive index

n20/D 1.393 (lit.)

bp

114-115 °C (lit.)

density

0.881 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; fruity; sweet

SMILES string

COC(=O)CC(C)C

InChI

1S/C6H12O2/c1-5(2)4-6(7)8-3/h5H,4H2,1-3H3

InChI key

OQAGVSWESNCJJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.9 °F - closed cup

Flash Point(C)

14.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service