Skip to Content
Merck
All Photos(1)

Key Documents

W253502

Sigma-Aldrich

Guaiacyl phenylacetate

≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2C6H4-2-(OCH3)
CAS Number:
Molecular Weight:
242.27
FEMA Number:
2535
EC Number:
Council of Europe no.:
238
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.711
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

mp

40-43 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

anise; spicy; chemical; phenolic; sweet

SMILES string

COc1ccccc1OC(=O)Cc2ccccc2

InChI

1S/C15H14O3/c1-17-13-9-5-6-10-14(13)18-15(16)11-12-7-3-2-4-8-12/h2-10H,11H2,1H3

InChI key

KNVYLIIBQXVDKC-UHFFFAOYSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service