Key Documents
SYNS321740
Synthware straight bore glass stopcock
bore size 4 mm, O.D. 10 mm stem, plug size 17/40
Synonym(s):
Synthware S321740, two way straigh bore stopcock
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S321740
bore size
4 mm
Related Categories
General description
Plugs and barrels are ground to be interchangeable, with 1:10 taper.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)