M46802
N-Methylformanilide
99%
Synonym(s):
N-Formyl-N-methylaniline, N-Methyl-N-phenylformamide
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
HCON(CH3)C6H5
CAS Number:
Molecular Weight:
135.16
Beilstein:
636496
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.561 (lit.)
bp
243-244 °C (lit.)
mp
8-13 °C (lit.)
density
1.095 g/mL at 25 °C (lit.)
SMILES string
CN(C=O)c1ccccc1
InChI
1S/C8H9NO/c1-9(7-10)8-5-3-2-4-6-8/h2-7H,1H3
InChI key
JIKUXBYRTXDNIY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
260.6 °F - closed cup
Flash Point(C)
127 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Mohamed Dribek et al.
Environmental science and pollution research international, 24(35), 27070-27076 (2014-08-12)
We demonstrated the use of a new organometallic nanoprobe for competitive surface-enhanced Raman scattering (SERS) immunoassay devoted to the detection of polycyclic aromatic hydrocarbons (PAH) such as benzo[a]pyrene (BaP) in seawater. The nanoprobes are gold nanoparticles (GNPs) labeled by a
Jisheng Zhang et al.
Dalton transactions (Cambridge, England : 2003), 44(21), 9847-9859 (2015-05-06)
Utilization of dioxygen as the terminal oxidant at ambient temperature is always a challenge in redox chemistry, because it is hard to oxidize a stable redox metal ion like iron(III) to its high oxidation state to initialize the catalytic cycle.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service