Skip to Content
Merck
All Photos(1)

Key Documents

L033000

Sigma-Aldrich

BTT Activator

0.3 M

Synonym(s):

5-(Benzylthio)-1H-tetrazole solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N4S
CAS Number:
Molecular Weight:
192.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

storage condition

dry at room temperature

concentration

51.9-63.4 g/L (BTT content, titration)

color

clear

SMILES string

C(Sc1nnn[nH]1)c2ccccc2

InChI

1S/C8H8N4S/c1-2-4-7(5-3-1)6-13-8-9-11-12-10-8/h1-5H,6H2,(H,9,10,11,12)

InChI key

GXGKKIPUFAHZIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Activator solution, containing powerful activators like 4,5-dicyanoimidazole(DCI), 5-(3,5-bis(trifluoromethyl)phenyl)-1H-tetrazole (Activator 42) or5-(Ethylthio)-1H-tetrazole (ETT) in acetonitrile, are mixed with solutions of phosphoramidites during the extension step. The activator reacts with the amidite group to form a highly reactive intermediate. The intermediate then forms an internucleotide bond with the detritylated 5′hydroxyl group of the growing oligonucleotide chain.BTT Activator 0.3M is 5-(Benzylthio)-1H-tetrazole.

Preparation Note

Acetonitrile / 5-(Benzylthio)-1H-Tetrazole 100 / 6 (v/w)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service