Skip to Content
Merck
All Photos(1)

Key Documents

JKA9013

Sigma-Aldrich

4arm-PEG2500-PLA3500

PEG average Mn 2,500, PCL average Mn 3,500

Synonym(s):

4arm-PEG-PLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(CH2O(CH2CH2O)n(COCHCH3O)mH)4
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

PCL average Mn 3,500
PEG average Mn 2,500

polymer architecture

shape: 4-arm
functionality: homofunctional

General description

4arm-PEG-PCL is a biodegradable material. It is especially used for drug delivery.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service