DWKBDA230P
Azlon Straight-sided Beakers
graduated, polypropylene, capacity 600 mL
Synonym(s):
BDA230P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polypropylene
feature
graduated
packaging
pkg of 10 ea
manufacturer/tradename
Azlon
storage condition
room temperature
capacity
600 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ideal for general laboratory use and management of liquid samples
Features and Benefits
Straight sided stackable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service