Key Documents
DWK233301504
DURAN® Pasteur pipette with short draw fine tip
glass, capacity (2 mL), pkg of 4 packs, pack of 250 ea, non-sterile
Synonym(s):
DURAN® Pasteur pipette with long draw fine tip
About This Item
UNSPSC Code:
41121500
NACRES:
NB.04
material
glass
sterility
non-sterile
feature
graduation: no
packaging
pack of 250 ea
pkg of 4 packs
manufacturer/tradename
DURAN®
capacity
(2 mL)
General description
Borosilicate glass 5.1 is more stable than soda-lime glass and is therefore insensitive to rapid temperature changes and chemical stress. Nearly inert borosilicate glass minimizes interactions with the material and reduces the risk of sample contamination. Pasteur pipettes with short or long extended, fine tip (150 and 230 mm). The filling volume is approx. 2 ml. Natural rubber teat can be ordered separately.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)