Skip to Content
Merck
All Photos(1)

Key Documents

DWK218656958

Duran® Gls 80 Laboratory Glass Bottle Protect

wide-neck, plastic-coated glass bottle (PU), with screw cap to drain contents (Without pouring ring), capacity 3500 mL

Synonym(s):

218656958

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43
capacity:
3500 mL
material:
plastic-coated glass bottle (PU)
wide-neck
feature:
with screw cap to drain contents (Without pouring ring)
packaging:
pkg of 1 ea

material

plastic-coated glass bottle (PU)
wide-neck

feature

with screw cap to drain contents (Without pouring ring)

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 218656958

capacity

3500 mL

Looking for similar products? Visit Product Comparison Guide

Application

Autoclaving of media and reagents, Preparation of media and reagents, Sample collection, Culturing vessel, Storage of media and reagents, Liquid handling reservoir, Boro 3.3, plastic coated

Features and Benefits

Easy access to the bottle contents due to the wide diameter of the bottle neck ; Optimum filling and pouring and particularly easy cleaning; Spoons, spatulas and tweezers can be manipulated with ease; Even large-volume funnels can be inserted into the bottles without any difficulty ; Stability due to the large base area; Easy-to-read scale with graduation marks to estimate the content; Fired-on, highly durable white ceramic; Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service