Key Documents
CLS5340250
Pyrex® heavy wall graduated filtering flask, with tubulation
10 mm O.D. hose-connection, capacity 250 mL
Synonym(s):
conical flasks, filtering flask, filtering flasks, flasks erlenmeyer, Corning® 5340
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
flat bottom flask
narrow-mouth
Joint
joint stopper6
sterility
non-sterile
feature
closure type Rubber stopper
packaging
case of 18
pack of 6
manufacturer/tradename
Corning 5340-250
volume measuring range
75-250 mL
capacity
250 mL
General description
These flasks have sidearm tabulations and are blown in special molds designed to give maximum mechanical strength. For convenience, the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)