CLS44501L
Pyrex® baffled Erlenmeyer flask
capacity 1,000 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, laboratory flasks, Corning® 4450
About This Item
Recommended Products
material
borosilicate glass
flat bottom flask
narrow-mouth
narrow-mouth neck
sterility
non-sterile
feature
cap: no
closure type Rubber stopper
packaging
case of 1
manufacturer/tradename
Corning 4450-1L
capacity
1,000 mL
General description
- Pyrex® baffled Erlenmeyer flasks feature a heavy tooled rim for maximum durability.
- Baffle-designed Delong flasks promote optimal and consistent solution agitation when used with flask shakers.
- Uniform wall thickness ensures an optimal balance between mechanical strength and thermal shock resistance.
- Includes a marking spot for convenience.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)