Skip to Content
Merck
All Photos(1)

Key Documents

CLS1500500

Corning® narrow mouth reagent bottle with Pyrex® ST stopper

capacity 500 mL, stopper joint: 24

Synonym(s):

glass bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

borosilicate glass bottle
clear bottle
cylindrical bottle
narrow-mouth bottle
polypropylene cap

Joint

joint: stopper24

feature

cap

packaging

pack of 1 ea
case of 24 ea

manufacturer/tradename

Corning 1500-500

bottle neck diam.

~35.5 mm

capacity

500 mL

diam. × H

86 mm × 163.5 mm

Looking for similar products? Visit Product Comparison Guide

General description

These Pyrex narrow mouth reagent storage bottles have sturdy ground necks with standard taper stoppers.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service