BR718020
BRAND® counting chamber BLAUBRAND® Thoma pattern
with clips, double ruled
Synonym(s):
BLAUBRAND®, Cell culture, Counting Chambers, Thoma
All Photos(1)
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Recommended Products
material
colorless
glass
sterility
non-sterile
feature
with clips
manufacturer/tradename
BRAND 718020
wells
1
General description
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.Rulings correspond to the central large square of the Neubauer chamber. The mini squares have an area of 0.0025 mm² each. Since the outer large squares are not completed, the Thoma chamber is only used for counting thrombocytes and erythrocytes.
Features and Benefits
- BLAUBRAND® quality
- Outstanding measurement precision
- 100% certified quality
Packaging
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)