Skip to Content
Merck
All Photos(1)

Key Documents

ADE000319

Sigma-Aldrich

2-Iodo-isonicotinic acid methyl ester

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6INO2
CAS Number:
Molecular Weight:
263.03
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Pricing and availability is not currently available.

form

solid

SMILES string

COC(=O)c1ccnc(I)c1

InChI

1S/C7H6INO2/c1-11-7(10)5-2-3-9-6(8)4-5/h2-4H,1H3

InChI key

XXOKMYMXDALOEN-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
1320450553900205
assay

≥98% β-phase basis (sintered Powder)

assay

≥98% β-phase basis (unsintered powder)

assay

-

assay

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

grade

puriss. p.a.

grade

puriss. p.a.

grade

-

grade

-

impurities

≤50 mg/kg total heavy metals as lead, ≤500 mg/kg total sulfur as SO4 (ICP)

impurities

≤250 mg/kg total sulfur (as SO4) (ICP), ≤50 mg/kg total heavy metals (as lead)

impurities

-

impurities

-

cation traces

As: ≤0.5 mg/kg, Cd: ≤1 mg/kg, Cr: ≤10 mg/kg, Hg: ≤0.5 mg/kg, Pb: ≤5 mg/kg, Ba: ≤20 mg/kg, Cu: ≤5 mg/kg, Zn: ≤20 mg/kg, Co: ≤1 mg/kg, Ni: ≤5 mg/kg

cation traces

As: ≤0.5 mg/kg, Ba: ≤10 mg/kg, Cd: ≤1 mg/kg, Co: ≤1 mg/kg, Cr: ≤5 mg/kg, Cu: ≤1 mg/kg, Hg: ≤0.5 mg/kg, Ni: ≤1 mg/kg, Pb: ≤5 mg/kg, Zn: ≤10 mg/kg

cation traces

-

cation traces

-

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of Adesis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    C W Somawardhana et al.
    International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes, 42(3), 215-220 (1991-01-01)
    The radiochemical syntheses of methyl 2-[123I]-iodoisonicotinate, 2-[123I]-iodoisonicotinic acid hydrazide and 2-[124I]-iodoisonicotinic acid hydrazide was accomplished. Iodine-123 was incorporated in the methyl ester molecule by an exchange reaction in glacial acetic acid. The average efficiency of iodine exchange reaction was (92.6

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service